missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid 37%, with Low Cadmium, Lead and Mercury Level, for Analysis, meets analytical specification of Ph.Eur., Fisher Chemical™

Catalog Number 12625167
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


36.74 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
500 mL
1 L
2.5 L
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
hydrogen chloride
Cl

Hydrochloric Acid 37%, with Low Cadmium, Lead and Mercury Level, for Analysis, meets analytical specification of Ph.Eur.

TRUSTED_SUSTAINABILITY
pH 1
Color Colorless
Melting Point -35°C
Boiling Point 57°C
Packaging HDPE plastic bottle
Vapor Pressure 125mbar at 20°C
Quantity 1 L
Viscosity 1.8 mPaS at 15°C
Physical Form Liquid
Formula Weight 36.46g/mol
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 37% with low Cadmium, Lead & Mercury level , for analysis Certified AR , Conform to EP
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'