missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 37%, Extra Pure, d=1.18, SLR, Fisher Chemical™

ClH, CAS Number-7647-01-0, hydrogen chloride hcl, muriatic acid, hydrochloric acid, chlorwasserstoff, hydrogen chloride, chlorohydric acid, chloorwaterstof, acide chlorhydrique, spirits of salt, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol

Brand:  Fisher Chemical H/1150/PB17

52.00 GBP valid until 2021-06-30
Use promo code "14804" to get your promotional price.



Code : 21

Additional Details : CAS Number : 7647-01-0 Weight : 2.95000kg Transport : UN number : 1789 Chem class : 8 Pack group : II


Packaging HDPE plastic bottle
Quantity 2.5L

 View more versions of this product

Product Code. 10000180

Quantity Price
1 £65.00 / 2.50 L
4 £57.00 / 2.50 L
16 £54.16 / 2.50 L
In stock, ready to ship: 432

Please to purchase this item. Need a web account? Register with us today!



Description

Description

ClH, CAS Number-7647-01-0, hydrogen chloride hcl, muriatic acid, hydrochloric acid, chlorwasserstoff, hydrogen chloride, chlorohydric acid, chloorwaterstof, acide chlorhydrique, spirits of salt, anhydrous hydrochloric acid, 2.5L, 57 deg.C, CHEBI:17883, Colorless, 36.46g/mol

Specifications

Chemical Identifiers

7647-01-0
36.458
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
chlorane
ClH
11324
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

1.8 mPaS at 15°C
2.5L
7647-01-0
11324
VEXZGXHMUGYJMC-UHFFFAOYSA-N
chlorane
313
36.46g/mol
HDPE plastic bottle
1.27
57°C
Colorless
1
ClH
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.458
CHEBI:17883
Liquid
125mbar at 20°C
-35°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Hydrochloric acid

Signal Word
  • Danger

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid

SDS Documents
View accessory products