missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™
107.35 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Poly Bottle
Specifications
Packaging | Narrow Mouth Poly Bottle |
Specific Gravity | 1L = 0.00kg |
Quantity | 1 L |
IUPAC Name | chlorane |
Formula Weight | 36.46 |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Hydrochloric Acid, 0.1N Volumetric Solution |
Safety and Handling
Signal Word
- Warning
- Substances and mixtures corrosive to metals Category 1
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P390-Absorb spillage to prevent material damage.
- P406-Store in corrosive resistant/ container with a resistant inner liner.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'