missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.1N Standardized Solution, Alfa Aesar™

Brand:  Alfa Aesar 035644.M4

Code : NEW

Additional Details : CAS Number : 7647-01-0 Weight : 0.01000kg

Quantity 20L

 View more versions of this product

Product Code. 11373777

  • £77.60 / 20 L


Estimated Shipment date
from Supplier 08-06-2018
Add to basket

Specifications

Chemical Identifiers

Hydrochloric acid
Liquid
36.46
7732-18-5
HCl
MFCD00011324

Specifications

Hydrochloric acid
Colorless liquid
HCl
MFCD00011324
23 hPa (17mm Hg) at 20°C
20L
7732-18-5
Liquid
36.46
Hydrochloric acid: 0.826%; Water: 99.174%
Clear colorless
Videos
SDS