missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid, 0.05N Standardized Solution, Thermo Scientific Chemicals

Catalog Number p-4781931
Click to view available options
Quantity:
1 L
4 L
20 L
This item is not returnable. View return policy
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
Cl
This item is not returnable. View return policy
7647-01-0, 7732-18-5
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
CHEBI:17883
Cl

Hydrochloric acid, 0.05N standardized solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 L
Color Colorless
IUPAC Name hydrogen chloride
Formula Weight 36.46
Concentration or Composition (by Analyte or Components) Hydrochloric acid: 0.413%; Water: 99.587%
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Miscible with water, methanol, ethanol and ether
Physical Form Liquid
Chemical Name or Material Hydrochloric acid, 0.05N

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'