missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.01N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391421
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Polyethylene Bottle
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
This item is not returnable. View return policy
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.00kg
Quantity 1 L
IUPAC Name chlorane
Formula Weight 36.46
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Hydrochloric Acid, 0.01N Volumetric Solution
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'