missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic Acid Solution, purum p.a., ≥62% (T), Honeywell™

Catalog Number 15641690
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br
This item is not returnable. View return policy
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
Br

Purum p.a.,≥62% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY
Color Brown-Yellow
Melting Point -11°C
Boiling Point 126°C
Density 1.73g/cu.cm
Packaging Glass Bottle
Quantity 250 mL
Physical Form Clear liquid
UN Number UN1788
Beilstein 3587158
IUPAC Name bromane
Formula Weight 80.91g/mol
Percent Purity ≥62% (T)
Grade purum p.a.
Chemical Name or Material Hydrobromic Acid Solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'