missing translation for 'onlineSavingsMsg'
Learn More

Hydrobromic acid, 48% w/w aq. soln., Thermo Scientific Chemicals

Catalog Number p-4408621
Click to view available options
Quantity:
940 mL
4 x 940 mL
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
hydrogen bromide
Br
10035-10-6
BrH
80.91
MFCD00011323
CPELXLSAUQHCOX-UHFFFAOYSA-N
hydrogen bromide, hydrobromic acid, bromwasserstoff, broomwaterstof, bromowodor, acido bromidrico, acide bromhydrique, anhydrous hydrobromic acid, bromowodor polish, hydrobromide
260
CHEBI:47266
hydrogen bromide
Br

Hydrobromic acid is an indispensable raw material for organic intermediates, inorganic pharmaceuticals, photosensitive materials, dyes and medicines. It is also used in the production of bromine compounds. It is used as an important agent in the industrial processes of isomerization, polymerization, hydration and dehydration and esterification reactions. It plays an essential role for the extraction of mercury from the cinnebar ore.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.49 g/mL
Sensitivity Air Sensitive
Quantity 940 mL
Physical Form Liquid
Beilstein 3587158
Solubility Information Miscible with water,alcohol and organic solvents.
Formula Weight 80.92
Chemical Name or Material Hydrobromic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'