missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrobromic acid, 99.9999% (metals basis), 48% w/w aq. soln., Thermo Scientific Chemicals
16.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Description
Hydrobromic acid is used for the production of inorganic bromides of zinc, calcium, and sodium. It is used as a catalyst in some alkylation reactions. In the field of metallurgy, it is employed in the extraction of certain ores. Further, it is used as a reagent for the cleavage of certain ether linkages or as a brominating agent in organic transformations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Appearance | Colorless to pale yellow liquid |
Color | Colorless to Yellow |
Density | 1.49 |
Sensitivity | Air Sensitive |
Vapor Pressure | 22500 hPa (16876mm Hg) |
Assay Percent Range | 99.9999% (metals basis) |
Quantity | 5 mL |
Physical Form | Liquid |
Beilstein | 3587158 |
Solubility Information | Miscible with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'