missing translation for 'onlineSavingsMsg'
Learn More

Hydriodic acid, for analysis, 57 wt.% aqueous solution, distilled, stabilized, Thermo Scientific Chemicals

Catalog Number p-7214340
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
50 mL
250 mL
1 L
10034-85-2
HI
127.91
MFCD00011347
XMBWDFGMSWQBCA-UHFFFAOYSA-N
hydrogen iodide
I
10034-85-2
HI
127.91
MFCD00011347
XMBWDFGMSWQBCA-UHFFFAOYSA-N
hydrogen iodide
I

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.7000 g/mL
Boiling Point 127.0°C
Packaging Glass bottle
Linear Formula HI
Quantity 50 mL
Fieser 01,449; 02,213; 06,281; 07,173; 08,246; 09,238; 10,199; 11,255; 15,166
Merck Index 15, 4814
Specific Gravity 1.7
Solubility Information Solubility aq. soln.: soluble.
Formula Weight 127.91
Grade Analysis
Chemical Name or Material Hydriodic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'