missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydridotetrakis(triphenylphosphine)rhodium(I), Rh 8.9% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
It finds its application as a catalyst for hydrogenation of alkenes and hydrosilylation of alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 145°C to 147°C (decomposition) |
Quantity | 1 g |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in chloroform, toluene |
IUPAC Name | rhodium;triphenylphosphane |
Formula Weight | 1153.09 |
Concentration or Composition (by Analyte or Components) | Rh 8.9% min |
Physical Form | Powder |
Chemical Name or Material | Hydridotetrakis (triphenylphosphine)rhodium (I) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'