missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog Number 10762592
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
5341-61-7
ClH5N2
68.50
MFCD00064543
BIVUUOPIAYRCAP-UHFFFAOYSA-N
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
17548
hydrazine;dihydrochloride
[H+].[Cl-].NN
5341-61-7
ClH5N2
68.50
MFCD00064543
BIVUUOPIAYRCAP-UHFFFAOYSA-N
hydrazine dihydrochloride, hydrazine, dihydrochloride, unii-km8dd9j0c8, ccris 5479, diamine dihydrochloride, hydrazinium monochloride, km8dd9j0c8, hydrazine, hydrochloride 1:2, hydrazinedihydrochloride, hydrazine hcl
17548
hydrazine;dihydrochloride
[H+].[Cl-].NN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 198.0°C
Color White
Physical Form Chunks and/or Powder
Quantity 500 g
Assay Percent Range 98.5% min. (Argentometry)
Linear Formula NH2NH2·2HCl
Merck Index 15, 4809
Solubility Information Solubility in water: soluble. Other solubilities: slightly soluble in alcohol
Formula Weight 104.97
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Hydrazine dihydrochloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'