Learn More
HYDRANAL™ - Solvent, Reagent for volumetric two-component KF titration (working medium), Honeywell Fluka™
Brand: Honeywell Fluka 34800-4X2.5L
Code : NEW
Additional Details : CAS Number : 288-32-4 Weight : 8.90000kg Transport : UN number : 1230 Chem class : 3 Pack group : II
Specifications
11°C (51.8°F) | |
HYDRANAL™-Solvent | |
MFCD00147197 |
4 x 2.5 L | |
0.89g/mL | |
UN3286 |
Safety and Handling
- HYDRANAL-Solvent
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 1B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H360D-May damage the unborn child.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol, imidazole,sulphur dioxide