missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™

Brand:  Honeywell Fluka 34741-1L

16.07 GBP valid until 2024-04-30
Use promo code "21741" to get your promotional price.



Code : NEW

Additional Details : CAS Number : 67-56-1 Weight : 0.79000kg Transport : UN number : 1230 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15644390

  • £16.92 / 1 L
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

9°C (48.2°F)
HYDRANAL™-Methanol dry
67-56-1
1 L
0.79g/mL
UN1230
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • HYDRANAL-Methanol dry

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS Documents

Documents

Certificates
Special Offers

Special Offers