missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution, For Ketones and Aldehydes, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 6283-01

Additional Details : Weight : 1.05000kg Transport : UN number : 1992 Chem class : 3 Pack group : III

Product Code. 15567634

Quantity Price
1 £375.34 / 500mL
6 £349.48 / 500mL
Estimated Shipment: 09-12-2021

Please to purchase this item. Need a web account? Register with us today!



Description

Description

    For Coulometric Karl Fischer Analysis
Specifications

Specifications

HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution
1L = 1.10kg
For Ketones and Aldehydes
BAKER ANALYZED™ Reagent
48.70°C
500mL
Narrow Mouth Amber Glass Bottle
Videos
SDS
Documents
Certificates