missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution, For Diaphragmless cell, Chloroform-Free, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 6285-01

527.25 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.



Additional Details : Weight : 0.95000kg Transport : UN number : 1993 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15597634

  • £555.00 / 500mL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

    For Coulometric Karl Fischer Analysis
Specifications

Specifications

46.00°C
HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution
Narrow Mouth Amber Glass Bottle
500 mL
For Diaphragmless cell, Chloroform-Free
BAKER ANALYZED™ Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers