missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution, For Diaphragmless cell, Chloroform-Free, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 6285-02

Additional Details : Weight : 1.71000kg Transport : UN number : 1993 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 15507644

  • £620.00 / 1 L
Estimated Shipment: 18-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

    For Coulometric Karl Fischer Analysis
Specifications

Specifications

46.00°C
HYDRA-POINT™ Karl Fischer Coulometric Vessel Solution
Narrow Mouth Amber Glass Bottle
1 L
For Diaphragmless cell, Chloroform-Free
BAKER ANALYZED™ Reagent
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P235-Store in a well-ventilated place. Keep cool.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers