missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydra-Point™ Coulometric Generator Solution, Universal, BAKER ANALYZED™ Reagent, J.T. Baker™
Click to view available options
Quantity:
5 mL
125 mL
Packaging:
Glass Ampoule
Narrow Mouth Amber Glass Bottle
Description
- Catholyte solution for coulometric analysis. For both general use and aldehydes and ketones.
Specifications
Flash Point | 46.00°C |
Packaging | Narrow Mouth Amber Glass Bottle |
Quantity | 125 mL |
Grade | BAKER ANALYZED™ Reagent |
Chemical Name or Material | Hydra-Point™ Coulometric Generator Solution, Universal |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H400-Very toxic to aquatic life.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'