missing translation for 'onlineSavingsMsg'
Learn More

HYDRA-POINT™ Buffer Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Brand:  JT Baker 6282-01

Additional Details : Weight : 1.05000kg Transport : UN number : 3286 Chem class : 3 Pack group : II

Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle

 View more versions of this product

Product Code. 15547634

  • £136.00 / 500mL


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

    For Karl Fischer titration
Specifications

Chemical Identifiers

HYDRA-POINT™ Buffer Solution
BAKER ANALYZED™ Reagent

Specifications

HYDRA-POINT™ Buffer Solution
500mL
BAKER ANALYZED™ Reagent
11.00°C
Narrow Mouth Amber Glass Bottle
Videos