missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Water for Injection (WFI) Quality Water
Click to view available options
Quantity:
500 mL
1 L
10 L
20 L
200 L
Description
- Format: Liquid
Specifications
IUPAC Name | water |
Molecular Weight (g/mol) | 18.02 |
Quantity | 20 L |
Chemical Name or Material | Water |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'