missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Hyclone™ Characterized Fetal Bovine Serum (Australia)

Characterized FBS irradiated is comparable to the highest quality FBS from other suppliers, but is lower priced than defined FBS.

Brand:  Cytiva SH30084.03HI

1068.21 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.



Code : A0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11950958

  • £1160.00 / 500mL
Estimated Shipment: 09-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Filtered through 3 sequential 0.1 μm pore size filters
Specifications

Specifications

AQIS
Australia
500 mL
Sequential Triple-filtered, 100 nm
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
HyClone™
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
Fetal Bovine Serum
Sterile
Heat Inactivated
Yes
USDA Inspected
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers