Learn More
HyClone products Cytiva Media; Cell Culture; HyClone; Liquid; HyQ SFM4MegaVir, w/Phenol red, w/o L-Glutamine; protein-free, animal-derived component free medium, designed for high virus yield in Vero, MDCK and MDBK cell lines; 500mL in PETE bottle
Brand:  HyClone products Cytiva SH30552.01
Media; Cell Culture; HyClone; Liquid; HyQ SFM4MegaVir, w/Phenol red, w/o L-Glutamine; protein-free, animal-derived component free medium, designed for high virus yield in Vero, MDCK and MDBK cell lines; 500mL in PETE bottle
Code : A0
Additional Details : Weight : 0.50000kg
Product Code. 10284732