missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ SFM4MAb™-Utility Media

Brand:  GE Healthcare Hyclone SH30382.02

Code : 83

Additional Details : Weight : 0.01250kg

 View more versions of this product

Product Code. 10133252

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Specifications

cGMP
Liquid
2° to 8°C
SFM4MAb-Utility Media, 1000mL
1000mL
L-Glutamine
Videos
Documentation