missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ PF-CHO™ LS and PF-CHO MPS Media

Support growth of multiple CHO cell clones and production of a variety of recombinant proteins. HyClone™ PF CHO™ LS and PF CHO MPS Media is a protein-free media developed through Metabolic Pathway Design.

Brand:  GE Healthcare Hyclone SH30220.01

Code : 83

Additional Details : Weight : 1.36500kg

 View more versions of this product

Product Code. 10390094

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Description

Description

  • Format: Liquid
Specifications

Specifications

cGMP
Liquid
2° to 8°C
HEPES
PF-CHO (liquid) - Protein free
1000mL
L-glutamine, sodium bicarbonate, phenol red
Videos