Learn More
Cytiva HyClone™ Cosmic Calf Serum (New Zealand origin)
A bovine calf serum fortified with iron and growth promoting factors for growing a broad spectrum of cell types.
Brand: Cytiva SH30413.02IR
Code : 85
Additional Details : Weight : 0.80000kg
Description
All New Zealand calf serum products are carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases. We follow the same standards in processing of our calf serum as with our fetal bovine serum (FBS) to enable offering the highest quality and traceability.Cosmic Calf Serum, New Zealand Origin is filtered through sequential 100 nm (0.1 µm) pore size-rated filters. Before dispensing, each lot of serum is pooled using true pool technology to ensure uniformity and consistency between bottles.
Specifications
500 mL | |
USDA Regulations | |
≤ 25 EU/mL | |
≤ 25 mg/dL | |
Gamma Irradiated | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C. | |
Gamma Irradiated Cosmic Calf Serum, New Zealand origin |
Sterile | |
Insulin,Gamma Globulin,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,pH,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Sodium,Potassium,Calcium,Chloride,Inorganic Phosphorus,Iron,TIBC,Percent Saturation,Glucose,IgG | |
No | |
Yes | |
Filtered through sequential 100 nm pore-size rated filters. | |
Yes | |
Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus |