missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ CDM4MAb™ Media

Brand:  GE Healthcare Hyclone SH30801.02

Code : 83

Additional Details : Weight : 0.01250kg

 View more versions of this product

Product Code. 10399663

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Specifications

With L-Glutamine
1000mL
L-Glutamine
Liquid
2° to 8°C
Videos
Documentation