missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Bovine Serum Albumin (BSA)
Support cell culture growth with HyClone™ Bovine Serum Albumin (BSA).
Brand: Cytiva SH30574.01
Code : 83
Additional Details : Weight : 0.01000kg
Description
- pH 7.0
- Lyophilized powder
Specifications
Packaged in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Bovine Serum Albumin (BSA), Cell Culture Grade, pH 7.0, Lyophilized Powder. 10g | |
Non-sterile | |
6.5 to 7.5 |
2° to 8° C | |
Powder | |
≤ 10 EU/mg | |
10 g |