missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ AdvanceSTEM™ Human Somatic Stem Cell Media

Brand:  GE Healthcare Hyclone SH30889.02

Code : A0

Additional Details : Weight : 0.45000kg

 View more versions of this product

Product Code. 10554274

  • £153.69 / 450mL


Estimated Shipment
12-06-2018
Add to basket

Specifications

Specifications

Mesenchymal Stem Cells
450mL
2° to 8°C
Chondrogenic Differentiation Medium, Replacement
Filter Sterilized - .2μm
Videos