missing translation for 'onlineSavingsMsg'
Learn More

HyClone™ AdvanceSTEM™ Amniotic Epithelial Expansion Kit

Brand:  HyClone™ SH30900.02

Code : A0

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10453125

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Alert:

Hyclone Cell Culture products now a part of GE Healthcare.


Description

Description

  • Includes a proprietary basal Amniotic Epithelial Expansion Medium, to which an aliquot of HyClone AdvanceSTEM Growth Supplement is added at the time of use. Basal Amniotic Epithelial Expansion Medium and Growth Supplement should be used together, but are available as separate components
Specifications

Specifications

Mesenchymal Stem Cells
AdvanceStem Amniotic Epithelial Growth Medium
Filter Sterilized - 0.2μm
SH3878.2, SH39.2
250mL
2° to 8°C
Videos