Learn More
BTX Molecular Delivery Systems™ Hybrimune™ Electrofusion System
Brand:  BTX Molecular Delivery Systems™ 470300N
Code : NEW
Additional Details : Weight : 10.69000kg
Product Code. 15447390
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
comprises generator, 2mL coaxial chambers, BTXpress™ Cytofusion Medium C, software and cables
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products