missing translation for 'onlineSavingsMsg'
Learn More

HX 630, Tocris Bioscience™

RXR synergist

Brand:  RD Systems (Bio-Techne) 3913/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13443804

  • £186.69 / 10mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RXR agonist. Acts as a retinoid synergist; enhances the potency of AM 80 (Cat. No. 3507) in a HL-60 cell differentiation assay.
Specifications

Chemical Identifiers

188844-52-2
441.589
4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl-benzoic acid, benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl, 4-7,7,10,10-tetramethyl-7,8,9,10-tetrahydro-5-thia-13-aza-benzo 4,5 cyclohepta 1,2-b naphthalen-12-yl-benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl benzoic acid, 4-14,14,17,17-tetramethyl-2-thia-9-azatetracyclo 9.8.0.0 3 ,?.0 1 3 , 1 ? nonadeca-1 19 ,3,5,7,9,11,13 18-heptaen-10-yl benzoic acid
4-(7,7,10,10-tetramethyl-8,9-dihydronaphtho[2,3-b][1,5]benzothiazepin-12-yl)benzoic acid
C28H27NO2S
PFGCWQPTOKPRRK-UHFFFAOYSA-N
9889522
CC1(CCC(C2=C1C=C3C(=C2)SC4=CC=CC=C4N=C3C5=CC=C(C=C5)C(=O)O)(C)C)C

Specifications

10mg
188844-52-2
4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl-benzoic acid, benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl, 4-7,7,10,10-tetramethyl-7,8,9,10-tetrahydro-5-thia-13-aza-benzo 4,5 cyclohepta 1,2-b naphthalen-12-yl-benzoic acid, 4-7,8,9,10-tetrahydro-7,7,10,10-tetramethylbenzo b naphtho 2,3-f 1,4 thiazepin-12-yl benzoic acid, 4-14,14,17,17-tetramethyl-2-thia-9-azatetracyclo 9.8.0.0 3 ,?.0 1 3 , 1 ? nonadeca-1 19 ,3,5,7,9,11,13 18-heptaen-10-yl benzoic acid
CC1(CCC(C2=C1C=C3C(=C2)SC4=CC=CC=C4N=C3C5=CC=C(C=C5)C(=O)O)(C)C)C
441.589
441.58
HX 630
C28H27NO2S
PFGCWQPTOKPRRK-UHFFFAOYSA-N
4-(7,7,10,10-tetramethyl-8,9-dihydronaphtho[2,3-b][1,5]benzothiazepin-12-yl)benzoic acid
9889522
>98%
Videos
Documents