missing translation for 'onlineSavingsMsg'
Learn More

HX 531, Tocris Bioscience™

Potent RXR antagonist

Brand:  Tocris (Bio-Techne) 391250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13433804

  • £946.68 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent RXR antagonist (IC50 = 18 nM). Promotes white and brown pre-adipocyte differentiation into white adipocytes. Also inhibits bexarotene -induced brown adipogenic reprogramming of myoblasts.
Specifications

Chemical Identifiers

188844-34-0
483.568
d02yhd, z-4-5,7,7,10,10-pentamethyl-2-nitro-7,8,9,10-tetrahydro-5h-benzo b naphtho 2,3-e 1,4 diazepin-12-yl benzoic acid, 4-5,7,7,10,10-pentamethyl-2-nitro-7,8,9,10-tetrahydro-5h-5,13-diaza-benzo 4,5 cyclohepta 1,2-b naphthalen-12-yl-benzoic acid, 4-5,7,7,10,10-pentamethyl-2-nitro-8,9-dihydronaphtho 2,3-b 1,5 benzodiazepin-12-yl benzoic acid, 4-7,8,9,10-tetrahydro-5,7,7,10.10-pentamethyl-2-nitro-5h-benzo b naphtho 2,3-e 1,4-diazepin-12-yl-benzoic acid
4-(5,7,7,10,10-pentamethyl-2-nitro-8,9-dihydronaphtho[2,3-b][1,5]benzodiazepin-12-yl)benzoic acid
C29H29N3O4
SXKPGYKPQPYJER-UHFFFAOYSA-N
11755040
CC1(CCC(C2=C1C=C3C(=C2)N(C4=C(C=C(C=C4)[N+](=O)[O-])N=C3C5=CC=C(C=C5)C(=O)O)C)(C)C)C

Specifications

50mg
188844-34-0
d02yhd, z-4-5,7,7,10,10-pentamethyl-2-nitro-7,8,9,10-tetrahydro-5h-benzo b naphtho 2,3-e 1,4 diazepin-12-yl benzoic acid, 4-5,7,7,10,10-pentamethyl-2-nitro-7,8,9,10-tetrahydro-5h-5,13-diaza-benzo 4,5 cyclohepta 1,2-b naphthalen-12-yl-benzoic acid, 4-5,7,7,10,10-pentamethyl-2-nitro-8,9-dihydronaphtho 2,3-b 1,5 benzodiazepin-12-yl benzoic acid, 4-7,8,9,10-tetrahydro-5,7,7,10.10-pentamethyl-2-nitro-5h-benzo b naphtho 2,3-e 1,4-diazepin-12-yl-benzoic acid
CC1(CCC(C2=C1C=C3C(=C2)N(C4=C(C=C(C=C4)[N+](=O)[O-])N=C3C5=CC=C(C=C5)C(=O)O)C)(C)C)C
483.568
483.56
HX 531
C29H29N3O4
SXKPGYKPQPYJER-UHFFFAOYSA-N
4-(5,7,7,10,10-pentamethyl-2-nitro-8,9-dihydronaphtho[2,3-b][1,5]benzodiazepin-12-yl)benzoic acid
11755040
>98%
Videos
Documents