missing translation for 'onlineSavingsMsg'
Learn More

Proteintech HumanKine™ recombinant human IL-5 protein

Product Code. 17640097 Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
1000 μg
Unit Size:
1000µg
100µg
10µg
This item is not returnable. View return policy

Product Code. 17640097

Brand: Proteintech HZ13241000UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

HumanKine™ recombinant protein

IL-5 is a 4-α-helix, pleiotropic cytokine that regulates the activities of B cells, eosinophils, and basophils. It is produced mostly by CD4+ T cells, eosinophils, and basophils, and signals through its specific receptor (IL-5R) on target cells to activate downstream JAK/STAT, Btk, and Ras/ERk pathways. Due to its pleiotropic effects, IL-5 plays a role in both innate and adaptive immune responses. It is a strong promoter of eosinophil differentiation, migration, maturation, and survival. Additionally, IL-5 is also capable of stimulating activated B cells to differentiate into antibody-secreting cells. Elevated IL-5 levels are strongly correlated with eosinophilia, which is a hallmark feature of allergy, asthma, and hypersensitivity (PMID: 21986312, 22174445, 19819937, 16895557)

Specifications

Accession Number P05113
Formulation 1 x PBS
Gene ID (Entrez) 3567
Molecular Weight (g/mol) 35-38 kDa
Quantity 1000 μg
Source HEK293
Storage Requirements Lyophilized product should be stored at -20°C to -80°C until expiry date and (at Room Temperature for 2 weeks.)
Endotoxin Concentration <0.1 EU/μg
Specific Reactivity Human
Recombinant Recombinant
Reconstitution Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein in sterile 1 xPBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA).
Activity 0.2-0.9 ng/mL EC50
Form Lyophilized
Purity or Quality Grade >95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.