missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WNT7B Partial ORF (NP_478679, 241 a.a. - 349 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£272.00 - £412.00
Specifications
Accession Number | NP_478679 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7477 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16187535
|
Abnova™
H00007477-Q01.25UG |
25 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16177535
|
Abnova™
H00007477-Q01.10UG |
10 ug |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene is a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Among members of the human WNT family, this gene product is most similar to WNT7A protein. [provided by RefSeq]
Sequence: VRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCKSpecifications
NP_478679 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WNT7B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
7477 | |
WNT7B (Human) Recombinant Protein (Q01) | |
VRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCK | |
RUO | |
WNT7B | |
Recombinant | |
wheat germ expression system |