Learn More
Abnova™ Human WNT3A Full-length ORF (AAI03922.1, 1 a.a. - 385 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00089780-P01.2ug
Additional Details : Weight : 0.00010kg
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 96% amino acid identity to mouse Wnt3A protein, and 84% to human WNT3 protein, another WNT gene product. This gene is clustered with WNT14 gene, another family member, in chromosome 1q42 region. [provided by RefSeq]
Sequence: MAPLGYFLLLCSLKQALGSYPIWWSLAVGPQYSSLGSQPILCASIPGLVPKQLRFCRNYVEIMPSVAEGIKIGIQECQHQFRGRRWNCTTVHDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTAAICGCSSRHQGSPGKGWKWGGCSEDIEFGGMVSREFADARENRPDARSAMNRHNNEAGRQAIASHMHLKCKCHGLSGSCEVKTCWWSQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRPRYTYFKVPTERDLVYYEASPNFCEPNPETGSFGTRDRTCNVSSHGIDGCDLLCCGRGHNARAERRREKCRCVFHWCCYVSCQECTRVYDVHTCKNPGSRAGNSAHQPPHPQPPVRFHPPLRRAGKVPSpecifications
AAI03922.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119418/MGC119419/MGC119420 | |
WNT3A | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
89780 | |
WNT3A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAPLGYFLLLCSLKQALGSYPIWWSLAVGPQYSSLGSQPILCASIPGLVPKQLRFCRNYVEIMPSVAEGIKIGIQECQHQFRGRRWNCTTVHDSLAIFGPVLDKATRESAFVHAIASAGVAFAVTRSCAEGTAAICGCSSRHQGSPGKGWKWGGCSEDIEFGGMVSREFADARENRPDARSAMNRHNNEAGRQAIASHMHLKCKCHGLSGSCEVKTCWWSQPDFRAIGDFLKDKYDSASEMVVEKHRESRGWVETLRPRYTYFKVPTERDLVYYEASPNFCEPNPETGSFGTRDRTCNVSSHGIDGCDLLCCGRGHNARAERRREKCRCVFHWCCYVSCQECTRVYDVHTCKNPGSRAGNSAHQPPHPQPPVRFHPPLRRAGKVP | |
RUO | |
WNT3A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |