missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human WDR90 Full-length ORF (AAH65836.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | AAH65836.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 197335 |
Molecular Weight (g/mol) | 58.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16176949
|
Abnova™
H00197335-P01.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16166949
|
Abnova™
H00197335-P01.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sequence: MLWFGQPGPGGWNRKCLRPFTLPRGQHPQPSYGFPQVNEVVFSPGESHCATCSEDGSVRVWALASMELVIQFQVLNQSCLCLAWSPPCCGRPEQQRLAAGYGDGSLRIFSVSRTAMELKMHPHPVALTTVAFSTDGQTVLSGDKDGLVAVSHPCTGTTFRVLSDHQGAPISTICVTCKETQGHLPPSLAAFCPWDGALLMYVGPGVYKEVIIYNLCQKQVVEKIPLPFFAMSLSLSPGTHLLAVGFAECMLRLVDCAMGTAQDFAGHDNAVHLCRFTPSARLLFTAARNEILVWEVPGLSpecifications
AAH65836.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MLWFGQPGPGGWNRKCLRPFTLPRGQHPQPSYGFPQVNEVVFSPGESHCATCSEDGSVRVWALASMELVIQFQVLNQSCLCLAWSPPCCGRPEQQRLAAGYGDGSLRIFSVSRTAMELKMHPHPVALTTVAFSTDGQTVLSGDKDGLVAVSHPCTGTTFRVLSDHQGAPISTICVTCKETQGHLPPSLAAFCPWDGALLMYVGPGVYKEVIIYNLCQKQVVEKIPLPFFAMSLSLSPGTHLLAVGFAECMLRLVDCAMGTAQDFAGHDNAVHLCRFTPSARLLFTAARNEILVWEVPGL | |
RUO | |
WDR90 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
197335 | |
WDR90 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C16orf15/C16orf16/C16orf17/C16orf18/C16orf19/FLJ36483/FLJ44660/KIAA1924 | |
WDR90 | |
Recombinant | |
wheat germ expression system |