missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VEGFR2/KDR Quantikine ELISA Kit

Human VEGFR2/KDR Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SVR200

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462896

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

The Human VEGFR2/KDR Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human VEGFR2/KDR/Flk-1 in cell culture supernates (100 ul), cell lysates (100 ul), serum (20 ul), edta plasma (20 ul), heparin plasma (20 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
6 Plates
Cell Culture Supernates (100uL),Cell Lysates (100uL),Serum (20uL),EDTA Plasma (20uL),Heparin Plasma (20uL)
VEGFR2/KDR/Flk-1
Serum
78.10 - 5000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3791
11.4pg/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
CD309, CD309 antigen, EC 2.7.10, EC 2.7.10.1, Fetal liver kinase 1, fetal liver kinase-1, Flk1, Flk-1, FLK1tyrosine kinase growth factor receptor, KDR, Kinase insert domain receptor, kinase insert domain receptor (a type III receptor tyrosine kinase), Protein-tyrosine kinase receptor flk-1, soluble VEGFR2, vascular endothelial growth factor receptor 2, VEGF R2, VEGFR, VEGFR2, VEGFR-2
Videos
Documents