missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human VEGF-A Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human VEGF-A.

Brand:  Invitrogen™ BMS277-2CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15521867

  • £359.99 / 96 tests


EU Stock 15
Add to basket

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human VEGF-A
Biotin-Conjugate anti-human VEGF-A polyclonal antibody
Streptavidin-HRP
Human VEGF-A Standard lyophilized, 2ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Vascular endothelial growth factor A, VPF
ELISA
7.9pg/mL
50μL
VEGF-A
Videos