missing translation for 'onlineSavingsMsg'
Learn More

Human Total ZAP70 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  RD Systems (Bio-Techne) DYC3709-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13454605

  • £338.10 / Pack of 2


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

  • Optimized capture and detection antibody pairings with recommended concentrations save lengthy development time
  • Development protocols are provided to guide further assay optimization
  • Assay can be customized to your specific needs
  • Economical alternative to complete kits
Specifications

Specifications

ELISA
Biotin
7535
2 Plates
1 Kit
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
Human
Videos
Documents