missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total XIAP DuoSet IC, R&D Systems™

Human Total XIAP DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC822-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434805

  • £751.17 / Pack of 5

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human Total XIAP DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human XIAP.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
331
5 Plates
Human
Videos
Documents