missing translation for 'onlineSavingsMsg'
Learn More

Human Total ROR2 DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC2064-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434435

  • £338.10 / Pack of 2

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
4920
2 Plates
Human
Videos
Documents