missing translation for 'onlineSavingsMsg'
Learn More

Human Total PON3 DuoSet IC, R&D Systems™

Brand:  RD Systems (Bio-Techne) DYC4345-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434675

  • £338.10 / Pack of 2

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Assay Type: ELISA Development Kit for intracellular proteins
Cross Reactivity: <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.

Specifications

Specifications

<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
5446
2 Plates
Human
Videos
Documents