missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total p21/CIP1/CDKN1A DuoSet IC, R&D Systems™

Human Total p21/CIP1/CDKN1A DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DYC1047-5

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13444265

  • £654.05 / Pack of 5

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human Total p21/CIP1/CDKN1A DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human p21/CIP1/CDKN1A.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
1026
5 Plates
Human
62.50 - 4000pg/mL
Videos