Learn More
R&D Systems™ Human Total MMP-7 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase ELISA designed to measure MMP-7 in cell culture supernates, serum, plasma, saliva, and urine.
Brand:  R&D Systems™ DMP700
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462695
  • £572.02 / 96 tests
Estimated Shipment
28-02-2019
Add to basket
Description
Description
It contains NS0-expressed recombinant human MMP-7 and antibodies raised against the recombinant factor. The Quantikine kit detected both pro- and active forms of recombinant human MMP-7. Natural human MMP-7 showed dose-response curves that were parallel to the standard curves obtained using the kit standards, indicating that this kit can be used to determine relative levels of natural human MMP-7
Specifications
Specifications
0.16 - 10ng/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
4316
1 Plate
0.094ng/mL
Cell Culture Supernates (25μL), Serum (25μL), Heparin Plasma (25μL), Saliva (25μL), Urine (50μL)
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (25uL), Serum (25uL), Heparin Plasma (25uL), Saliva (25uL), Urine (50uL)