missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total MMP-7 Quantikine ELISA Kit
Human Total MMP-7 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.16 to 10 ng/mL |
---|---|
Assay Sensitivity | 0.094 ng/mL |
Sample Type | Cell Culture Supernatants, Serum, Plasma (Heparin), Saliva, Urine, Plasma (Heparin), Saliva, Serum, Urine |
Description
The Human Total MMP-7 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-7 in cell culture supernates (25 ul), serum (25 ul), heparin plasma (25 ul), saliva (25 ul), urine (50 ul).Specifications
0.16 to 10 ng/mL | |
0.094 ng/mL | |
Cell Culture Supernatants, Serum, Plasma (Heparin), Saliva, Urine, Plasma (Heparin), Saliva, Serum, Urine |