missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total MMP-3 Quantikine ELISA Kit, R&D Systems™

Human Total MMP-3 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DMP300

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452695

  • £621.81 / 96 tests


Estimated Shipment date
from Supplier 25-03-2020
Add to basket

Description

Description

The Human Total MMP-3 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-3 in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul).
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.045ng/mL
Store the unopened product at 2° to 8°C. Do not use past expiration date.
Serum
0.16 - 10ng/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
4314
1 Plate
Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents