Learn More
R&D Systems™ Human Total MMP-3 Quantikine ELISA Kit, R&D Systems™
a 4.5 hour solid phase immunoassay designed to measure total MMP-3 (pro- and active MMP-3) in cell culture supernates, serum, and plasma.
Brand:  R&D Systems™ SMP300
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13462826
  • £3037.79 / Pack of 6
Estimated Shipment
03-04-2019
Add to basket
Description
Description
It contains NS0-expressed recombinant human Pro-MMP-3 and antibodies raised against the recombinant factor. Both antibodies also recognize recombinant human active MMP-3. Natural human MMP-3 showed dose-response curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards, indicating that this kit can be used to determine relative levels of natural human MMP-3
Specifications
Specifications
0.16 - 10ng/mL
Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
4314
6 Plates
0.045ng/mL
Cell Culture Supernates (50μL), Serum (10μL), Heparin Plasma (10μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Serum (10uL), Heparin Plasma (10uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos