missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total Mer DuoSet IC, R&D Systems™
Human Total Mer DuoSet IC ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Product Type | ELISA Development Kit for intracellular proteins |
Quantity | 1 Kit |
Gene ID (Entrez) | 10461 |
Species | Human |
Description
Complete ELISpot kits are highly sensitive, microplate-based assays for the detection of cytokine secreting cells. Kits are available for detection and enumeration of a single analyte or two analytes simultaneously. Complete ELISpot kits are ready-to-r
The Human Total Mer DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human Mer.Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
1 Kit | |
Human |
ELISA Development Kit for intracellular proteins | |
10461 |