missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Total MDM2/HDM2 DuoSet IC, R&D Systems™

contains the basic components required for the development of sandwich ELISAs to measure in cell lysates

Brand:  R&D Systems™ DYC1244-2

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13434295

  • £427.77 / Pack of 2

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human Total MDM2/HDM2 DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human MDM2/HDM2.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit for intracellular proteins
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
4193
2 Plates
Human
Videos
Documents