missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Total IkB-alpha DuoSet IC, R&D Systems™
contains the basic components required for the development of sandwich ELISAs to measure in cell lysates
POA
Specifications
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
---|---|
Product Type | ELISA Development Kit for intracellular proteins |
Quantity | 1 Kit |
Gene ID (Entrez) | 4792 |
Species | Human |
Description
The Human Total IkB-alpha DuoSet IC ELISA is a Solid Phase Sandwich ELISA that quantifies human IkB-alpha.Specifications
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
1 Kit | |
Human |
ELISA Development Kit for intracellular proteins | |
4792 |